Product Details

SNP ID
rs1065782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:45891959 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCTGCCGAATAGAACTCTACAGA[A/G]TCGTAGAGAGTTTAGCCAAGGCACA
Phenotype
MIM: 146730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGFBP1 PubMed Links

Gene Details

Gene
IGFBP1
Gene Name
insulin like growth factor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000596.2 840 Missense Mutation ATC,GTC I183V NP_000587.1

View Full Product Details