Product Details

SNP ID
rs909281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144840984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGTTTCCTCGCCAGCTCGGATG[C/G]TGGAGCCCCTGACCCCGCAGCCCAA
Phenotype
MIM: 608216 MIM: 194531
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COMMD5 PubMed Links

Gene Details

Gene
COMMD5
Gene Name
COMM domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081003.2 Intron NP_001074472.1
NM_001081004.2 Intron NP_001074473.1
NM_001287237.1 Intron NP_001274166.1
NM_014066.3 Intron NP_054785.2
Gene
ZNF7
Gene Name
zinc finger protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282795.1 Intron NP_001269724.1
NM_001282796.1 Intron NP_001269725.1
NM_001282797.1 Intron NP_001269726.1
NM_003416.3 Intron NP_003407.1
XM_006716654.2 Intron XP_006716717.1
XM_006716656.3 Intron XP_006716719.1
XM_011517291.2 Intron XP_011515593.1
XM_011517292.2 Intron XP_011515594.1
XM_011517293.2 Intron XP_011515595.1
XM_011517294.2 Intron XP_011515596.1
XM_011517295.2 Intron XP_011515597.1
XM_011517296.2 Intron XP_011515598.1
XM_011517297.2 Intron XP_011515599.1
XM_017013813.1 Intron XP_016869302.1
XM_017013814.1 Intron XP_016869303.1
XM_017013815.1 Intron XP_016869304.1
XM_017013816.1 Intron XP_016869305.1
XM_017013817.1 Intron XP_016869306.1

View Full Product Details