Product Details

SNP ID
rs140344040
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:72916289 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GCACAAGAGGGAGCCAAGATACAGT[G/C]CCTGGTTGATCTGAGGCAAGGATCC
Phenotype
MIM: 300878
Polymorphism
G/C, Transversion Substitution
Allele Nomenclature
Literature Links
DMRTC1 PubMed Links

Gene Details

Gene
DMRTC1
Gene Name
DMRT like family C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033053.2 Intron NP_149042.2
XM_005262287.3 Intron XP_005262344.1
XM_005262288.3 Intron XP_005262345.1
XM_017029725.1 Intron XP_016885214.1

View Full Product Details