Product Details

SNP ID
rs1048531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128458017 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTACCCCGGGGTCTGGTGTATGCT[A/G]TGCTTTCCTCAGCAGTATGGCTCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HILPDA PubMed Links

Gene Details

Gene
HILPDA
Gene Name
hypoxia inducible lipid droplet associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098786.1 897 UTR 3 NP_001092256.1
NM_013332.3 897 UTR 3 NP_037464.1

View Full Product Details