Product Details

SNP ID
rs965691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:155079932 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGAAATTCTCTTACATAACCATG[A/G]TACATTTATAAAAATTGGGAAATTA
Phenotype
MIM: 300617 MIM: 300116
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRCC3 PubMed Links

Gene Details

Gene
BRCC3
Gene Name
BRCA1/BRCA2-containing complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018055.2 Intron NP_001018065.1
NM_001242640.1 Intron NP_001229569.1
NM_024332.3 Intron NP_077308.1
XM_005274751.4 Intron XP_005274808.1
XM_017029838.1 Intron XP_016885327.1
Gene
CMC4
Gene Name
C-X9-C motif containing 4
There are no transcripts associated with this gene.

Gene
MTCP1
Gene Name
mature T-cell proliferation 1
There are no transcripts associated with this gene.

View Full Product Details