Product Details

SNP ID
rs937114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56473902 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGGTAAATAAATAGACACGTAAT[G/T]TTTAAAGTAATTGTGATAAATAGTG
Phenotype
MIM: 606365
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GLS2 PubMed Links

Gene Details

Gene
GLS2
Gene Name
glutaminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280796.1 Intron NP_001267725.1
NM_001280797.1 Intron NP_001267726.1
NM_001280798.1 Intron NP_001267727.1
NM_013267.3 Intron NP_037399.2
XM_005268797.1 Intron XP_005268854.1
XM_017019180.1 Intron XP_016874669.1
Gene
SPRYD4
Gene Name
SPRY domain containing 4
There are no transcripts associated with this gene.

View Full Product Details