Product Details

SNP ID
rs705703
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55997702 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTACCTGCACTGCTCTGGTCTAGTA[C/T]AAACAGGCTGCTGGCATTGAGGTAG
Phenotype
MIM: 606239 MIM: 179514 MIM: 606887
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
IKZF4 PubMed Links

Gene Details

Gene
IKZF4
Gene Name
IKAROS family zinc finger 4
There are no transcripts associated with this gene.

Gene
RAB5B
Gene Name
RAB5B, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
SUOX
Gene Name
sulfite oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000456.2 652 UTR 5 NP_000447.2
NM_001032386.1 652 UTR 5 NP_001027558.1
NM_001032387.1 652 Intron NP_001027559.1
XM_005269112.1 652 UTR 5 XP_005269169.1
XM_017019905.1 652 UTR 5 XP_016875394.1
XM_017019906.1 652 Intron XP_016875395.1
XM_017019907.1 652 UTR 5 XP_016875396.1
XM_017019908.1 652 Intron XP_016875397.1

View Full Product Details