Product Details

SNP ID
rs773113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55982866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATTCTAACTCCCACATTTTCCT[G/T]TATTGTAGAGTAGTAACATTGAGGC
Phenotype
MIM: 179514
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAB5B PubMed Links

Gene Details

Gene
RAB5B
Gene Name
RAB5B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252036.1 Intron NP_001238965.1
NM_001252037.1 Intron NP_001238966.1
NM_002868.3 Intron NP_002859.1
XM_005269051.2 Intron XP_005269108.1
XM_005269052.2 Intron XP_005269109.1

View Full Product Details