Product Details

SNP ID
rs1801152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:102840473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAATCCTTTGGGTGTATGGGTC[G/A]TAGCGAACTGAGAAGGGCCGAGGTA
Phenotype
MIM: 612349
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
PAH PubMed Links

Gene Details

Gene
PAH
Gene Name
phenylalanine hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000277.1 1714 Silent Mutation TAC,TAT Y414Y NP_000268.1
XM_017019370.1 1714 Intron XP_016874859.1

View Full Product Details