Product Details

SNP ID
rs944145
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:167432322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGAATGCCTGTCCCCCTGGTCC[A/G]GGGGCCAGGAAGGAGAAGGATCAAC
Phenotype
MIM: 186780 MIM: 164175
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD247 PubMed Links

Gene Details

Gene
CD247
Gene Name
CD247 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000734.3 Intron NP_000725.1
NM_198053.2 Intron NP_932170.1
XM_011510144.2 Intron XP_011508446.1
XM_011510145.1 Intron XP_011508447.1
XM_017002800.1 Intron XP_016858289.1
XM_017002801.1 Intron XP_016858290.1
Gene
POU2F1
Gene Name
POU class 2 homeobox 1
There are no transcripts associated with this gene.

View Full Product Details