Product Details

SNP ID
rs1135787
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:161122156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGAGGGAATAGGTCAGGGCAA[A/T]GCTTGCAGCATCAAGTTCCTCAGGA
Phenotype
MIM: 606841 MIM: 604618 MIM: 613466
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DEDD PubMed Links

Gene Details

Gene
DEDD
Gene Name
death effector domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039711.1 1184 Silent Mutation GCA,GCT A316A NP_001034800.1
NM_001039712.1 1184 Silent Mutation GCA,GCT A316A NP_001034801.1
NM_032998.2 1184 Silent Mutation GCA,GCT A316A NP_127491.1
XM_005245597.3 1184 Silent Mutation GCA,GCT A346A XP_005245654.1
XM_005245598.4 1184 Silent Mutation GCA,GCT A346A XP_005245655.1
XM_005245599.2 1184 Silent Mutation GCA,GCT A346A XP_005245656.1
XM_005245600.2 1184 Silent Mutation GCA,GCT A346A XP_005245657.1
XM_017002802.1 1184 Silent Mutation GCA,GCT A316A XP_016858291.1
Gene
NIT1
Gene Name
nitrilase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185092.1 1184 Intron NP_001172021.1
NM_001185093.1 1184 Intron NP_001172022.1
NM_001185094.1 1184 Intron NP_001172023.1
NM_005600.2 1184 Intron NP_005591.1
XM_005245214.2 1184 Intron XP_005245271.1
XM_005245215.4 1184 Intron XP_005245272.1
XM_005245216.3 1184 Intron XP_005245273.1
XM_006711341.3 1184 Intron XP_006711404.1
XM_017001371.1 1184 Intron XP_016856860.1
Gene
PFDN2
Gene Name
prefoldin subunit 2
There are no transcripts associated with this gene.

View Full Product Details