Product Details

SNP ID
rs947421
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:69567534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGCCGGCAGCCGCGCTCCGCCC[C/G]GCGCTGAGATTCGTGCCACCTGCCT
Phenotype
MIM: 614453
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LRRC7 PubMed Links

Gene Details

Gene
LRRC7
Gene Name
leucine rich repeat containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020794.2 405 Intron NP_065845.1
XM_017001885.1 405 Intron XP_016857374.1
XM_017001886.1 405 Intron XP_016857375.1
XM_017001887.1 405 Intron XP_016857376.1
XM_017001888.1 405 Intron XP_016857377.1
XM_017001889.1 405 Intron XP_016857378.1
XM_017001890.1 405 UTR 5 XP_016857379.1
XM_017001891.1 405 UTR 5 XP_016857380.1
XM_017001892.1 405 Intron XP_016857381.1
XM_017001893.1 405 Intron XP_016857382.1
XM_017001894.1 405 Intron XP_016857383.1
XM_017001895.1 405 Intron XP_016857384.1

View Full Product Details