Product Details

SNP ID
rs202674
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:49207449 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTTAAACCCATCTGGTCCATGCCCT[A/G]TTCTTCTAAATCCCGAAAGAGGGTC
Phenotype
MIM: 600934
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOLH1 PubMed Links

Gene Details

Gene
FOLH1
Gene Name
folate hydrolase (prostate-specific membrane antigen) 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014986.1 Intron NP_001014986.1
NM_001193471.1 Intron NP_001180400.1
NM_001193472.1 Intron NP_001180401.1
NM_001193473.1 Intron NP_001180402.1
NM_004476.1 Intron NP_004467.1
XM_011519958.2 Intron XP_011518260.2
XM_017017432.1 Intron XP_016872921.1
XM_017017433.1 Intron XP_016872922.1
XM_017017434.1 Intron XP_016872923.1
XM_017017435.1 Intron XP_016872924.1
XM_017017436.1 Intron XP_016872925.1
XM_017017437.1 Intron XP_016872926.1
XM_017017438.1 Intron XP_016872927.1
XM_017017439.1 Intron XP_016872928.1
XM_017017440.1 Intron XP_016872929.1
XM_017017441.1 Intron XP_016872930.1
XM_017017442.1 Intron XP_016872931.1
XM_017017443.1 Intron XP_016872932.1
XM_017017444.1 Intron XP_016872933.1
XM_017017445.1 Intron XP_016872934.1
XM_017017446.1 Intron XP_016872935.1
XM_017017447.1 Intron XP_016872936.1
XM_017017448.1 Intron XP_016872937.1
XM_017017449.1 Intron XP_016872938.1
XM_017017450.1 Intron XP_016872939.1
XM_017017451.1 Intron XP_016872940.1

View Full Product Details