Product Details

SNP ID
rs709688
Assay Type
Validated
NCBI dbSNP Submissions
75
Location
Chr.1:40787513 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGAACCTGCATCTTTCTGGCTCCAA[A/G]AGGTGGGAGATTGCCTAGAGTACAA
Phenotype
MIM: 603537
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNQ4 PubMed Links

Gene Details

Gene
KCNQ4
Gene Name
potassium voltage-gated channel subfamily Q member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004700.3 Intron NP_004691.2
NM_172163.2 Intron NP_751895.1
XM_017002792.1 Intron XP_016858281.1

View Full Product Details