Product Details

SNP ID
rs12541126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22053720 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCCCTGCCTGAGAGATACAGGAA[C/T]CCTGGCCCCGTGTTGTGGACACAAG
Phenotype
MIM: 125305 MIM: 603725
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DMTN PubMed Links

Gene Details

Gene
DMTN
Gene Name
dematin actin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114135.4 151 Intron NP_001107607.1
NM_001114136.2 151 Intron NP_001107608.1
NM_001114137.3 151 UTR 5 NP_001107609.1
NM_001114138.2 151 Intron NP_001107610.1
NM_001114139.3 151 Intron NP_001107611.1
NM_001302816.2 151 Intron NP_001289745.1
NM_001302817.2 151 Intron NP_001289746.1
NM_001323378.1 151 Intron NP_001310307.1
NM_001323380.1 151 Intron NP_001310309.1
NM_001323381.1 151 Intron NP_001310310.1
NM_001323382.1 151 Intron NP_001310311.1
NM_001323383.1 151 Intron NP_001310312.1
NM_001323384.1 151 Intron NP_001310313.1
NM_001323385.1 151 Intron NP_001310314.1
NM_001323387.1 151 Intron NP_001310316.1
NM_001323388.1 151 Intron NP_001310317.1
NM_001323389.1 151 Intron NP_001310318.1
NM_001323390.1 151 Intron NP_001310319.1
NM_001323391.1 151 Intron NP_001310320.1
NM_001323392.1 151 Intron NP_001310321.1
NM_001323393.1 151 Intron NP_001310322.1
NM_001323394.1 151 Intron NP_001310323.1
NM_001323395.1 151 Intron NP_001310324.1
NM_001323396.1 151 Intron NP_001310325.1
NM_001323397.1 151 Intron NP_001310326.1
NM_001323398.1 151 Intron NP_001310327.1
NM_001323399.1 151 Intron NP_001310328.1
NM_001323400.1 151 Intron NP_001310329.1
NM_001323401.1 151 Intron NP_001310330.1
NM_001978.4 151 Intron NP_001969.2
XM_005273432.1 151 UTR 5 XP_005273489.1
XM_005273433.1 151 Intron XP_005273490.1
XM_005273442.1 151 Intron XP_005273499.1
XM_011544435.1 151 Intron XP_011542737.1
XM_017013189.1 151 Intron XP_016868678.1
XM_017013190.1 151 Intron XP_016868679.1
XM_017013191.1 151 Intron XP_016868680.1
XM_017013192.1 151 Intron XP_016868681.1
XM_017013193.1 151 Intron XP_016868682.1
XM_017013194.1 151 Intron XP_016868683.1
XM_017013195.1 151 Intron XP_016868684.1
XM_017013196.1 151 Intron XP_016868685.1
XM_017013197.1 151 Intron XP_016868686.1
XM_017013198.1 151 Intron XP_016868687.1
Gene
FGF17
Gene Name
fibroblast growth factor 17
There are no transcripts associated with this gene.

View Full Product Details