Product Details

SNP ID
rs1146646
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:75799815 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGATAGATGGTAGTAGTATTTTCT[A/G]ATTCTGGGAATTTTGTAGGCAAAGC
Phenotype
MIM: 602105 MIM: 179080
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MSH4 PubMed Links

Gene Details

Gene
MSH4
Gene Name
mutS homolog 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002440.3 Intron NP_002431.2
Gene
RABGGTB
Gene Name
Rab geranylgeranyltransferase beta subunit
There are no transcripts associated with this gene.

View Full Product Details