Product Details

SNP ID
rs6122438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63401318 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGTCAGCCCCCACCCTTACAAGAC[A/G]GCAAAGTTCAGCAAACTGGAACCCC
Phenotype
MIM: 602235
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNQ2 PubMed Links

Gene Details

Gene
KCNQ2
Gene Name
potassium voltage-gated channel subfamily Q member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004518.5 7565 UTR 3 NP_004509.2
NM_172106.2 7565 UTR 3 NP_742104.1
NM_172107.3 7565 UTR 3 NP_742105.1
NM_172108.4 7565 UTR 3 NP_742106.1
NM_172109.2 7565 Intron NP_742107.1
XM_011528810.1 7565 UTR 3 XP_011527112.1
XM_011528811.1 7565 UTR 3 XP_011527113.1
XM_017027841.1 7565 UTR 3 XP_016883330.1
XM_017027842.1 7565 UTR 3 XP_016883331.1
XM_017027843.1 7565 UTR 3 XP_016883332.1
XM_017027844.1 7565 UTR 3 XP_016883333.1
XM_017027845.1 7565 UTR 3 XP_016883334.1
Gene
LOC100130152
Gene Name
uncharacterized LOC100130152
There are no transcripts associated with this gene.

View Full Product Details