Product Details

SNP ID
rs1784029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66332516 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCCCCCTGGGTCTCAGGCCACTC[C/T]GCCCGGCGGTAGACGAGGTAGCCAG
Phenotype
MIM: 606259 MIM: 605965
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRMS1 PubMed Links

Gene Details

Gene
BRMS1
Gene Name
breast cancer metastasis suppressor 1
There are no transcripts associated with this gene.

Gene
RIN1
Gene Name
Ras and Rab interactor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004292.2 2588 Silent Mutation GCA,GCG A704A NP_004283.2
XM_011545399.2 2588 Silent Mutation GCA,GCG A676A XP_011543701.1
XM_011545400.2 2588 Silent Mutation GCA,GCG A642A XP_011543702.1
XM_011545401.2 2588 Silent Mutation GCA,GCG A623A XP_011543703.1
XM_017018587.1 2588 Silent Mutation GCA,GCG A643A XP_016874076.1

View Full Product Details