Product Details

SNP ID
rs35494108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88458671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACCAACCCCATGGACCCGGCCC[G/T]GCCACACTCGGGGCCCTGGTGGGGC
Phenotype
MIM: 601950
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZFPM1 PubMed Links

Gene Details

Gene
ZFPM1
Gene Name
zinc finger protein, FOG family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153813.2 Intron NP_722520.2
XM_011522912.2 Intron XP_011521214.1
XM_011522914.2 Intron XP_011521216.1
XM_011522917.2 Intron XP_011521219.1
XM_017022982.1 Intron XP_016878471.1

View Full Product Details