Product Details

SNP ID
rs1055938
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:100196157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCCTTAATCATACGATACAAATC[A/G]TTTACAATATAAATTGTAAAGATGA
Phenotype
MIM: 248610
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DBT PubMed Links

Gene Details

Gene
DBT
Gene Name
dihydrolipoamide branched chain transacylase E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001918.3 1807 UTR 3 NP_001909.3
XM_005270545.3 1807 UTR 3 XP_005270602.1
XM_017000468.1 1807 UTR 3 XP_016855957.1
XM_017000469.1 1807 UTR 3 XP_016855958.1

View Full Product Details