Product Details

SNP ID
rs1866973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:153873607 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAGCTTTGGGGACTGAGGTATGC[C/G]TGCCTTAAACTACCATGCATTCATT
Phenotype
MIM: 608369
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GALNT13 PubMed Links

Gene Details

Gene
GALNT13
Gene Name
polypeptide N-acetylgalactosaminyltransferase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301627.1 Intron NP_001288556.1
NM_052917.3 Intron NP_443149.2
XM_011510537.2 Intron XP_011508839.1
XM_011510538.2 Intron XP_011508840.1
XM_017003258.1 Intron XP_016858747.1
XM_017003259.1 Intron XP_016858748.1
XM_017003260.1 Intron XP_016858749.1
XM_017003261.1 Intron XP_016858750.1
XM_017003262.1 Intron XP_016858751.1
XM_017003263.1 Intron XP_016858752.1

View Full Product Details