Product Details
- SNP ID
-
rs13000209
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:150470348 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACACATTGCAAAACGTTTGCTGATA[C/T]GTTAACTGAATTTTCCAAGTGTCAT
- Phenotype
-
MIM: 602924
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RND3
PubMed Links
Gene Details
- Gene
- RND3
- Gene Name
- Rho family GTPase 3
View Full Product Details