Product Details

SNP ID
rs7108225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4907611 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTCCCTCTCCTCCCTTCCTACCA[C/T]GTTGAGGGTCTTCTTGTTCAATGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR51A7 PubMed Links

Gene Details

Gene
OR51A7
Gene Name
olfactory receptor family 51 subfamily A member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004749.1 242 Missense Mutation ACG,ATG T81M NP_001004749.1
Gene
OR51G2
Gene Name
olfactory receptor family 51 subfamily G member 2
There are no transcripts associated with this gene.

View Full Product Details