Product Details

SNP ID
rs4713139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28124907 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGCAGAGGCATCTGTCTGATCCC[C/T]TTCCAGGGTTGGCACACATATCCTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN16 PubMed Links

Gene Details

Gene
ZSCAN16
Gene Name
zinc finger and SCAN domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320555.1 Intron NP_001307484.1
NM_001320556.1 Intron NP_001307485.1
NM_001320557.1 Intron NP_001307486.1
NM_001320558.1 Intron NP_001307487.1
NM_025231.2 Intron NP_079507.1
XM_017011324.1 Intron XP_016866813.1
Gene
ZSCAN16-AS1
Gene Name
ZSCAN16 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details