Product Details

SNP ID
rs34296786
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:137968198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCAGGAAAAATTATAGTAAATA[A/C]CTATAAATAGTAAAGTTGGTTATTG
Phenotype
MIM: 605238
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
HNMT PubMed Links

Gene Details

Gene
HNMT
Gene Name
histamine N-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024074.2 Intron NP_001019245.1
NM_001024075.1 Intron NP_001019246.1
NM_006895.2 Intron NP_008826.1
XM_011511064.2 Intron XP_011509366.1
XM_017003948.1 Intron XP_016859437.1
XM_017003949.1 Intron XP_016859438.1

View Full Product Details