Product Details
- SNP ID
-
rs1558868
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:32350999 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AAGGAAACAAATTGTTCCCCCATTG[A/G]TCTCCTCTTTAATGACTTACAGCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF507
PubMed Links
Gene Details
- Gene
- ZNF507
- Gene Name
- zinc finger protein 507
View Full Product Details