Product Details
- SNP ID
-
rs11672345
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:41403814 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGACTGGCTCCTGTCCAAGGCCGC[A/G]TGGCTGGAAAGAGACAGAACCAGAT
- Phenotype
-
MIM: 608348
MIM: 606492
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BCKDHA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114855860] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BCKDHA
- Gene Name
- branched chain keto acid dehydrogenase E1, alpha polypeptide
- Gene
- EXOSC5
- Gene Name
- exosome component 5
There are no transcripts associated with this gene.
View Full Product Details