Product Details

SNP ID
rs2279524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116512483 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATATCCACTGGATTGTCCACAT[A/G]TAAATACTTAAGTACTTTAAAACAG
Phenotype
MIM: 614137
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM26E PubMed Links

Gene Details

Gene
FAM26E
Gene Name
family with sequence similarity 26 member E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153711.3 Intron NP_714922.1
Gene
TRAPPC3L
Gene Name
trafficking protein particle complex 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139444.2 Intron NP_001132916.1

View Full Product Details