Product Details

SNP ID
rs3788368
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:24182018 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCCACCCCATTTCCCATCGACCCTC[C/T]TGCCACTTATCTCACTTGCAAGCAT
Phenotype
MIM: 604251 MIM: 615825
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CABIN1 PubMed Links

Gene Details

Gene
CABIN1
Gene Name
calcineurin binding protein 1
There are no transcripts associated with this gene.

Gene
SUSD2
Gene Name
sushi domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019601.3 Intron NP_062547.1

View Full Product Details