Product Details
- SNP ID
-
rs12960225
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:35291292 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGGCGGGGTGGGGAGGATGAGGG[A/G]GTGTCACTCTATGCTAGTGTCTTCA
- Phenotype
-
MIM: 604754
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF271P
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114365446] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF271P
- Gene Name
- zinc finger protein 271, pseudogene
There are no transcripts associated with this gene.
- Gene
- ZSCAN30
- Gene Name
- zinc finger and SCAN domain containing 30
There are no transcripts associated with this gene.
View Full Product Details