Product Details
- SNP ID
-
rs2760
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:79905758 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AAACCTGGACACCAACCAACAGAAT[A/C]CTCCCGTCCTTTGAAATTTCCATTA
- Phenotype
-
MIM: 608041
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ANTXR2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113322831] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANTXR2
- Gene Name
- anthrax toxin receptor 2
View Full Product Details