Product Details

SNP ID
rs1711841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:92052577 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAACTCTGCTGATACTGGATGGT[A/G]GTTGAGAGCATGGGCTTTGGAATAA
Phenotype
MIM: 610606
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPEB3 PubMed Links

Gene Details

Gene
CPEB3
Gene Name
cytoplasmic polyadenylation element binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178137.1 Intron NP_001171608.1
NM_014912.4 Intron NP_055727.3
XM_005269630.4 Intron XP_005269687.1
XM_006717714.3 Intron XP_006717777.1
XM_006717715.3 Intron XP_006717778.1
XM_011539514.2 Intron XP_011537816.1
XM_011539515.2 Intron XP_011537817.1
XM_011539517.2 Intron XP_011537819.1
XM_011539518.2 Intron XP_011537820.1
XM_011539519.2 Intron XP_011537821.1
XM_011539520.2 Intron XP_011537822.1
XM_011539522.2 Intron XP_011537824.1
XM_011539523.2 Intron XP_011537825.1
XM_017015927.1 Intron XP_016871416.1

View Full Product Details