Product Details

SNP ID
rs968497
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:200872982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAGCAAACCCCCTCCTGCTCA[A/G]AGCTGCTGCCGCCTGCGCCCAGGGC
Phenotype
MIM: 602174
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR25 PubMed Links

Gene Details

Gene
GPR25
Gene Name
G protein-coupled receptor 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005298.3 28 UTR 5 NP_005289.2

View Full Product Details