Product Details

SNP ID
rs1047247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:80266949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCTGCCACTGATAGGAACCCTA[G/T]AGGCTCCAGCAGCAGACAGAGCAGC
Phenotype
MIM: 165190
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FGF5 PubMed Links

Gene Details

Gene
FGF5
Gene Name
fibroblast growth factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291812.1 362 Intron NP_001278741.1
NM_004464.3 362 Missense Mutation AGA,ATA R42I NP_004455.2
NM_033143.2 362 Missense Mutation AGA,ATA R42I NP_149134.1

View Full Product Details