Product Details

SNP ID
rs1176819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:74975619 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAACAGACTAACTTATTAGATTGT[A/C]AATAGGGTAAACTCAAATAGCAGAC
Phenotype
MIM: 616552
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C9orf41-AS1 PubMed Links

Gene Details

Gene
C9orf41-AS1
Gene Name
C9orf41 antisense RNA 1
There are no transcripts associated with this gene.

Gene
CARNMT1
Gene Name
carnosine N-methyltransferase 1
There are no transcripts associated with this gene.

View Full Product Details