Product Details
- SNP ID
-
rs1039128
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
68
- Location
-
Chr.1:233620319 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCAGAATGATACTTGAAAGGTACGC[C/T]AAGAGTATTCTGAATATTTCACTTT
- Phenotype
-
MIM: 601745
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KCNK1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72762235] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCNK1
- Gene Name
- potassium two pore domain channel subfamily K member 1
- Gene
- MIR4427
- Gene Name
- microRNA 4427
There are no transcripts associated with this gene.
View Full Product Details