Product Details

SNP ID
rs2734582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31535767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATGCTTTATGGGACCTTCTCCC[A/G]ACCCTTTCCTGCCCAAGCCCCAGCC
Phenotype
MIM: 606853 MIM: 142560 MIM: 609624
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V1G2 PubMed Links

Gene Details

Gene
ATP6V1G2
Gene Name
ATPase H+ transporting V1 subunit G2
There are no transcripts associated with this gene.

Gene
ATP6V1G2-DDX39B
Gene Name
ATP6V1G2-DDX39B readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
DDX39B
Gene Name
DEAD-box helicase 39B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004640.6 Intron NP_004631.1
NM_080598.5 Intron NP_542165.1
Gene
DDX39B-AS1
Gene Name
DDX39B antisense RNA 1
There are no transcripts associated with this gene.

Gene
MCCD1
Gene Name
mitochondrial coiled-coil domain 1
There are no transcripts associated with this gene.

Gene
SNORD117
Gene Name
small nucleolar RNA, C/D box 117
There are no transcripts associated with this gene.

Gene
SNORD84
Gene Name
small nucleolar RNA, C/D box 84
There are no transcripts associated with this gene.

View Full Product Details