Product Details

SNP ID
rs1218825
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27435783 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATATGCCTTCTCCTCATTTTTGCT[C/G]AAAGCACGAAGAACACACATTAAAG
Phenotype
MIM: 600860
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GTF3A PubMed Links

Gene Details

Gene
GTF3A
Gene Name
general transcription factor IIIA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002097.2 1617 Intron NP_002088.2
Gene
MTIF3
Gene Name
mitochondrial translational initiation factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166261.1 1617 Intron NP_001159733.1
NM_001166262.1 1617 Intron NP_001159734.1
NM_001166263.1 1617 Missense Mutation TTC,TTG F243L NP_001159735.1
NM_152912.4 1617 Intron NP_690876.3
XM_006719771.3 1617 Missense Mutation TTC,TTG F243L XP_006719834.1
XM_006719772.3 1617 Missense Mutation TTC,TTG F243L XP_006719835.1
XM_011534957.2 1617 Missense Mutation TTC,TTG F243L XP_011533259.1
XM_011534958.2 1617 Missense Mutation TTC,TTG F243L XP_011533260.1
XM_011534959.2 1617 Missense Mutation TTC,TTG F243L XP_011533261.1
XM_011534960.2 1617 Missense Mutation TTC,TTG F243L XP_011533262.1
XM_011534961.2 1617 Missense Mutation TTC,TTG F243L XP_011533263.1
XM_011534962.2 1617 Missense Mutation TTC,TTG F243L XP_011533264.1
XM_011534963.2 1617 Missense Mutation TTC,TTG F243L XP_011533265.1
XM_017020417.1 1617 Intron XP_016875906.1

View Full Product Details