Product Details
- SNP ID
-
rs1800033
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:2164325 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCACGGCCTGGGGGCTGTCCAGCA[C/T]GTCGATGGCCAGCGTGTACGGGTCG
- Phenotype
-
MIM: 176730
MIM: 191290
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
INS
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3842724] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- INS
- Gene Name
- insulin
There are no transcripts associated with this gene.
- Gene
- INS-IGF2
- Gene Name
- INS-IGF2 readthrough
There are no transcripts associated with this gene.
- Gene
- MIR4686
- Gene Name
- microRNA 4686
There are no transcripts associated with this gene.
- Gene
- TH
- Gene Name
- tyrosine hydroxylase
View Full Product Details