Product Details
- SNP ID
-
rs1065687
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:2131409 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTAGATATGCTTATTGTTTTCATC[C/T]AATTTTGTGGGGGTGTGCGTGTGTG
- Phenotype
-
MIM: 147470
MIM: 610146
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IGF2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79531557] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IGF2
- Gene Name
- insulin like growth factor 2
- Gene
- IGF2-AS
- Gene Name
- IGF2 antisense RNA
There are no transcripts associated with this gene.
- Gene
- INS-IGF2
- Gene Name
- INS-IGF2 readthrough
- Gene
- MIR483
- Gene Name
- microRNA 483
There are no transcripts associated with this gene.
View Full Product Details