Product Details

SNP ID
rs1717770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1949893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCCGGCAGTGGCAGGGGCGGGAA[C/T]GGGGGTGGGGGACGGTAGGACGGTG
Phenotype
MIM: 600789
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL23 PubMed Links

Gene Details

Gene
MRPL23
Gene Name
mitochondrial ribosomal protein L23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021134.3 Intron NP_066957.3
XM_006718271.2 Intron XP_006718334.1
XM_011520273.1 Intron XP_011518575.1
XM_011520275.2 Intron XP_011518577.1
XM_011520276.2 Intron XP_011518578.1
Gene
SNORD131
Gene Name
small nucleolar RNA, C/D box 131
There are no transcripts associated with this gene.

View Full Product Details