Product Details
- SNP ID
-
rs947535
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
52
- Location
-
Chr.1:145728795 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCTCCTTGACATTAAGGAAAGGGT[G/A]AAGAATATATTGTCTTTTTCTTTTT
- Phenotype
-
MIM: 604463
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD160
PubMed Links
Gene Details
- Gene
- CD160
- Gene Name
- CD160 molecule
View Full Product Details