Product Details

SNP ID
rs944770
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:206512294 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTTTCAAAGAGGCCTTCTGAGCT[C/T]CCCACGCTGCAGCCCGTTTTTCCTC
Phenotype
MIM: 607020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RASSF5 PubMed Links

Gene Details

Gene
RASSF5
Gene Name
Ras association domain family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182663.3 Intron NP_872604.1
NM_182664.3 Intron NP_872605.1
NM_182665.3 Intron NP_872606.1

View Full Product Details