Product Details
- SNP ID
-
rs1473937
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:3627015 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CAAAATTTGGAGGGTAAGTGTATAT[G/T]TTTCTGCAGAGAGGGTCTACAACTT
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TRPC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7109228,rs7123007] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TRPC2
- Gene Name
- transient receptor potential cation channel subfamily C member 2, pseudogene
There are no transcripts associated with this gene.
View Full Product Details