Product Details

SNP ID
rs1749864
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:117063579 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTATCAGTAATTCATTCATTTTT[A/C]TTGTGGAGTTGAATTTCATTTATAA
Phenotype
MIM: 604718
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929099 PubMed Links

Gene Details

Gene
LOC101929099
Gene Name
uncharacterized LOC101929099
There are no transcripts associated with this gene.

Gene
TTF2
Gene Name
transcription termination factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003594.3 Intron NP_003585.3
XM_005271277.3 Intron XP_005271334.1
XM_011542303.2 Intron XP_011540605.1
XM_017002549.1 Intron XP_016858038.1
XM_017002550.1 Intron XP_016858039.1
XM_017002551.1 Intron XP_016858040.1
XM_017002552.1 Intron XP_016858041.1
XM_017002553.1 Intron XP_016858042.1
XM_017002554.1 Intron XP_016858043.1
XM_017002555.1 Intron XP_016858044.1

View Full Product Details