Product Details

SNP ID
rs1064631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:48076111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTACAATATGAATTGATATTATCT[C/T]TGGGTAGAATGCTAACAATTTTATT
Phenotype
MIM: 605718
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED4 PubMed Links

Gene Details

Gene
MED4
Gene Name
mediator complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270629.1 1904 UTR 3 NP_001257558.1
NM_014166.3 1904 UTR 3 NP_054885.1
XM_017020549.1 1904 UTR 3 XP_016876038.1
Gene
MED4-AS1
Gene Name
MED4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details