Product Details
- SNP ID
-
rs1246072
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:231742713 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTTGTGATCAGACTCTCTTCCCAAA[C/G]AAGAGAGTCTCTACCTTGTGGAAGG
- Phenotype
-
MIM: 602676
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PDE6D
PubMed Links
Gene Details
- Gene
- PDE6D
- Gene Name
- phosphodiesterase 6D
View Full Product Details