Product Details

SNP ID
rs932603
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:119507112 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTATTTTTCTGAGATAAGGATCCCA[C/T]AGGAGGAGAGAGCAATGAGTACATG
Phenotype
MIM: 109715
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSD3B1 PubMed Links

Gene Details

Gene
HSD3B1
Gene Name
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000862.2 1 Intron NP_000853.1
XM_011541314.2 1 UTR 5 XP_011539616.1
XM_017001119.1 1 Intron XP_016856608.1

View Full Product Details