Product Details
- SNP ID
-
rs1054607
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:109493208 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTTCCATAAGAGTGCTCCTTCCTGG[C/T]TGGCAGACCTTTGAGTTCACCTGAA
- Phenotype
-
MIM: 615358
MIM: 614064
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AK9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17602109] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AK9
- Gene Name
- adenylate kinase 9
- Gene
- ZBTB24
- Gene Name
- zinc finger and BTB domain containing 24
There are no transcripts associated with this gene.
View Full Product Details