Product Details
- SNP ID
-
rs1780584
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:151295045 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAACAGAAGCAAGCAGGCTGAGTG[C/G]AGCTGAAATGTAAAATAGCGTGGGG
- Phenotype
-
MIM: 602758
MIM: 610568
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PI4KB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113758311] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PI4KB
- Gene Name
- phosphatidylinositol 4-kinase beta
- Gene
- ZNF687
- Gene Name
- zinc finger protein 687
There are no transcripts associated with this gene.
View Full Product Details